missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Trichostatin A

Product Code. 17140052
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 17140052

Brand: STEMCELL Technologies 72282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epigenetic modifier; Inhibits histone deacetylase (HDAC)1 and HDAC6

  • Trichostatin A is a potent and reversible inhibitor of Histone Deacetylase (HDAC), therefore acting as an epigenetic modifier by preventing the removal of acetyl groups from lysine residues on histone tails
  • Trichostatin A inhibits both class I and class II HDACs, including HDAC1 (IC50 = 6 nM), HDAC4 (IC50 = 38 nM), and HDAC6 (IC50 = 8.6 nM)
  • (Furumai et al.; Yoshida et al.)

Specifications

CAS 58880-19-6
Synonym TSA
Quantity 1 mg
Molecular Formula C17H22N2O3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.