missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichomonas Vaginalis 1 Antibody (1306/156), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-62964CL1
This item is not returnable.
View return policy
Description
Trichomonas Vaginalis 1 Monoclonal antibody specifically detects Trichomonas Vaginalis 1 in Protozoa samples. It is validated for ELISA
Specifications
| Trichomonas Vaginalis 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| T vaginalis, TV | |
| Native Trichomonas vaginalis | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA | |
| 1306/156 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction