missing translation for 'onlineSavingsMsg'
Learn More

Trichloromethyl chloroformate, 98%

Product Code. 11468037
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468037 250 g 250g
11448037 10 g 10g
11458037 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468037 Supplier Thermo Scientific Alfa Aesar Supplier No. A17444.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 503-38-8 | C2Cl4O2 | 197.82 g/mol

Trichloromethyl chloroformate is used as a reagent in the synthesis of organic compounds. It serves as a source of phosgene used in some laboratory preparations. Also, it is used as a reactant for the synthesis of cyclic carbamimidates, N-alkenyl and cycloalkyl carbamates and prostate-specific membrane antigen-targeted anticancer prodrugs. In addition, it is involved in the preparation of an erythromycin A derivatives and antibody-drug conjugates. It is utilized in the conversion of amines, carboxylic acids, formamides in to isocyanates, acid chlorides and isocyanides respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloromethyl chloroformate is used as a reagent in the synthesis of organic compounds. It serves as a source of phosgene used in some laboratory preparations. Also, it is used as a reactant for the synthesis of cyclic carbamimidates, N-alkenyl and cycloalkyl carbamates and prostate-specific membrane antigen-targeted anticancer prodrugs. In addition, it is involved in the preparation of an erythromycin A derivatives and antibody-drug conjugates. It is utilized in the conversion of amines, carboxylic acids, formamides in to isocyanates, acid chlorides and isocyanides respectively.

Solubility
Miscible with ethanol, ether and benzene. Immiscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comStore in a cool place. Light and heat sensitive. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503-38-8
Molecular Formula C2Cl4O2
Molecular Weight (g/mol) 197.82
MDL Number MFCD00015553
InChI Key HCUYBXPSSCRKRF-UHFFFAOYSA-N
Synonym diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
PubChem CID 10426
IUPAC Name trichloromethyl carbonochloridate
SMILES ClC(=O)OC(Cl)(Cl)Cl

Specifications

Melting Point -57°C
Density 1.64
Boiling Point 128°C
Flash Point >110°C (230°F)
Assay Percent Range 98%
Odor Pungent
Linear Formula ClCO2CCl3
Refractive Index 1.457
Quantity 250 g
UN Number UN3390
Beilstein 970225
Sensitivity Moisture sensitive
Merck Index 14,3334
Solubility Information Miscible with ethanol,ether and benzene. Immiscible with water.
Formula Weight 197.83
Percent Purity 98%
Chemical Name or Material Trichloromethyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.