missing translation for 'onlineSavingsMsg'
Learn More

Trichloroisocyanuric acid, 90+%

Product Code. 11416766
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416766 500 g 500g
11406766 100 g 100g
11426766 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416766 Supplier Thermo Scientific Alfa Aesar Supplier No. B23906.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.

Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).

Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-90-1
Molecular Formula C3Cl3N3O3
Molecular Weight (g/mol) 232.40
MDL Number MFCD00006553
InChI Key YRIZYWQGELRKNT-UHFFFAOYSA-N
Synonym trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal
PubChem CID 6909
ChEBI CHEBI:33015
SMILES ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O

Specifications

Melting Point ∼245°C (decomposition)
Density 2.07
pH 3
Flash Point >250°C (482°F)
Odor Pungent
Quantity 500 g
UN Number UN2468
Beilstein 202022
Sensitivity Moisture sensitive
Merck Index 14,9641
Solubility Information Soluble in chlorocarbon,acetonitrile,acetone,water (12g/L).
IUPAC Name trichloro-1,3,5-triazinane-2,4,6-trione
Formula Weight 232.41
Percent Purity ≥90%
Chemical Name or Material Trichloroisocyanuric acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroisocyanuric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Symclosene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.