missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetyl Isocyanate, TRC

Product Code. 30456235
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456235 Supplier Toronto Research Chemicals Supplier No. TRCT7739051G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trichloroacetyl Isocyanate
CAS 3019-71-4
Molecular Formula C3Cl3NO2
Quantity 1 g
Synonym 2,2,2-Trichloro-acetyl Isocyanate, Trichloro-acetic Acid Anhydride with Isocyanic Acid, Trichloro-acetyl Isocyanate, Isocyanic Acid Anhydride with Trichloroacetic Acid, 2,2,2-Trichloroacetyl Isocyanate, Trichloroacetyl Isocyanate, alpha,alpha,alpha-Trichloroacetyl Isocyanate
InChI Formula InChI=1S/C3Cl3NO2/c4-3(5,6)2(9)7-1-8
SMILES ClC(Cl)(Cl)C(=O)N=C=O
IUPAC Name 2,2,2-trichloroacetyl isocyanate
Molecular Weight (g/mol) 188.4
Formula Weight 186.8995
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.