missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic anhydride, 95%

Product Code. 11334324
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334324 50 g 50g
11344324 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334324 Supplier Thermo Scientific Alfa Aesar Supplier No. L00849.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4124-31-6 | C4Cl6O3 | 308.741 g/mol

Trichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.

Solubility
Miscible with ether and acetic acid.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-31-6
Molecular Formula C4Cl6O3
Molecular Weight (g/mol) 308.741
MDL Number MFCD00000793
InChI Key MEFKFJOEVLUFAY-UHFFFAOYSA-N
Synonym trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
PubChem CID 20079
IUPAC Name (2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl

Specifications

Density 1.69
Boiling Point 98°C to 100°C (11 mmHg)
Refractive Index 1.484
Quantity 50 g
UN Number UN3265
Beilstein 980355
Sensitivity Moisture sensitive
Solubility Information Miscible with ether and acetic acid.
Formula Weight 308.76
Percent Purity 95%
Chemical Name or Material Trichloroacetic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.