missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
Description
Trichloroacetic Acid, Extra Pure, SLR
Specifications
pH | 1.2 |
Melting Point | 52°C |
Boiling Point | 196°C |
Color | White |
Formula Weight | 163.39g/mol |
Quantity | 500 g |
Packaging | Plastic powder jar |
Physical Form | Solid |
Vapor Pressure | 1.2mbar at 50°C, 0.08mbar @25C |
Safety and Handling
Product Identifier
- Trichloroacetic acid
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'