missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™

Catalog Number p-7073624
61.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


74.77 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
250 g
500 g
1 kg
76-03-9
C2HCl3O2
163.378
4177
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
6421
CHEBI:30956
2,2,2-trichloroacetic acid
C(=O)(C(Cl)(Cl)Cl)O
76-03-9
C2HCl3O2
163.378
4177
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
6421
CHEBI:30956
2,2,2-trichloroacetic acid
C(=O)(C(Cl)(Cl)Cl)O

C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol

TRUSTED_SUSTAINABILITY
pH 1.2
Melting Point 52°C
Boiling Point 196°C
Color White
Formula Weight 163.39g/mol
Quantity 250 g
Packaging Plastic powder jar
Physical Form Solid
Vapor Pressure 1.2mbar at 50°C, 0.08mbar @25C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'