missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid 30% Solution For Laboratory Use, Chem Lab

Product Code. 15908132
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15908132 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15908132 Supplier Chem-Lab Supplier No. CL02.2018.1000
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 76-03-9
Melting Point 0°C
Boiling Point 100°C
Color Colorless
Density 1.13 g/mL
Quantity 1 L
Packaging Glass bottles
Physical Form Liquid
Molecular Formula C2HCl3O2
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES OC(=O)C(Cl)(Cl)Cl
IUPAC Name trichloroacetic acid
Molecular Weight (g/mol) 163.38
Show More Show Less
compliance-icons compliance-icons
Identificativo prodotto
  • Trichloroacetic acid 30% w/v solution
Avvertenza
  • Danger
Categoria di pericolo
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Consigli di prudenza
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Informazioni supplementari
  • MIXTURE LIST-Contain: Trichloroacetic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.