missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid 100 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer

Product Code. 30312807
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312807 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312807 Supplier Dr. Ehrenstorfer Supplier No. DREA17683500MB100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trichloroacetic acid (TCA)
CAS 76-03-9
Analyte or Component Names Trichloroacetic acid (TCA)
Shipping Condition +20°C
Molecular Formula C2 H Cl3 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Trichloroacetic acid, 2,2,2-Trichloroacetic acid
InChI Formula InChI=1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
SMILES OC(=O)C(Cl)(Cl)Cl
IUPAC Name 2,2,2-trichloroacetic acid
Solvent or Matrix MTBE
Molecular Weight (g/mol) 163.39
Quantity 1 mL
Formula Weight 161.9042 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.