missing translation for 'onlineSavingsMsg'
Learn More

Tricarballylic acid, 99%

Product Code. 10018831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10018831 50 g 50g
10391004 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10018831 Supplier Thermo Scientific Acros Supplier No. 139360500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-14-9 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-14-9
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00002723
InChI Key KQTIIICEAUMSDG-UHFFFAOYSA-N
Synonym tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid
PubChem CID 14925
ChEBI CHEBI:45969
IUPAC Name propane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(CC(O)=O)C(O)=O

Specifications

Melting Point 154°C to 159°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO2CCH2CH(CO2H)CH2CO2H
Quantity 50 g
Beilstein 02, 815
Merck Index 15, 9787
Formula Weight 176.13
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tricarballylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tricarballylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.