missing translation for 'onlineSavingsMsg'
Learn More

Tricaprilin, MedChemExpress

Product Code. 16438816
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16438816

Brand: Medchem Express HYB1804

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Tricaprilin (Trioctanoin) is used in study for patients with mild to moderate Alzheimer's disease and has a role as an anticonvulsant and a plant metabolite.

Tricaprilin (Trioctanoin) is used in study for patients with mild to moderate Alzheimer's disease and has a role as an anticonvulsant and a plant metabolite.

Specifications

Chemical Name or Material Tricaprilin
CAS 538-23-8
Color Colorless
Purity Grade Notes Research
Molecular Formula C27H50O6
Quantity 100 mg
Synonym Trioctanoin Glyceryl trioctanoate
Solubility Information DMSO : 41.67 mg/mL (88.53 mM; Need ultrasonic)
SMILES CCCCCCCC(OCC(OC(CCCCCCC)=O)COC(CCCCCCC)=O)=O
Molecular Weight (g/mol) 470.68
Formula Weight 470.68
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.