missing translation for 'onlineSavingsMsg'
Learn More

Tricaine Methanesulfonate, Mikromol™

Product Code. 17971556
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
17971556 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 17971556

Brand: Mikromol MM3272.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tricaine Methanesulfonate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tricaine Methanesulfonate
CAS 886-86-2
Analyte or Component Names Tricaine Methanesulfonate
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O2 . C H4 O3 S
Certifications/Compliance ISO 17034
Synonym Benzoic acid, 3-amino-, ethyl ester, methanesulfonate (1:1), Benzoic acid, 3-amino-, ethyl ester, methanesulfonate (9CI), Benzoic acid, m-amino-, ethyl ester, methanesulfonate (7CI,8CI), Ethyl 3-aminobenzoate methanesulfonate, Ethyl m-aminobenzoate methanesulfonate, Finquel, MS 222, Metacain, Metacaine, NSC 93790, TS 222, Tricaine, Tricaine methanesulfonate, Benzocaine Impurity C as Methanesulfonate
InChI Formula InChI=1S/C9H11NO2.CH4O3S/c1-2-12-9(11)7-4-3-5-8(10)6-7;1-5(2,3)4/h3-6H,2,10H2,1H3;1H3,(H,2,3,4)
SMILES CCOC(=O)c1cccc(N)c1.CS(=O)(=O)O
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
Molecular Weight (g/mol) 261.29
Quantity 250 mg
Formula Weight 261.0671 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.