missing translation for 'onlineSavingsMsg'
Learn More

Tributyrin, 98.4%, MP Biomedicals™

Product Code. 11413511 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413511 250 mL 250mL
11403511 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413511 Supplier MP Biomedicals Supplier No. 0210311183
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C15H26O6
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Color Colorless
Density 1.032 g/mL
Quantity 250 mL
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
IUPAC Name 1,3-bis(butanoyloxy)propan-2-yl butanoate
Molecular Weight (g/mol) 302.37
PubChem CID 6119
ChEBI CHEBI:27971
Formula Weight 302.36
Physical Form Liquid
Chemical Name or Material Tributyrin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.