missing translation for 'onlineSavingsMsg'
Learn More

Tributylhexadecylphosphonium Bromide, TRC

Product Code. 30457016
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457016 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457016 Supplier Toronto Research Chemicals Supplier No. TRCT895990250MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tributylhexadecylphosphonium Bromide
CAS 14937-45-2
Molecular Formula C28H60BrP
Quantity 250 mg
Synonym Phosphonium','tributylhexadecyl-','bromide (8CI,9CI), Cetyltributylphosphonium bromide, Cyphos IL 162, Hexadecyltributylphosphonium bromide, PX 416B, TBHDPB, Tributylcetylphosphonium bromide
InChI Formula InChI=1S/C28H60P.BrH/c1-5-9-13-14-15-16-17-18-19-20-21-22-23-24-28-29(25-10-6-2,26-11-7-3)27-12-8-4;/h5-28H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
IUPAC Name tributyl(hexadecyl)phosphanium;bromide
Molecular Weight (g/mol) 507.65
Formula Weight 506.3616
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.