missing translation for 'onlineSavingsMsg'
Learn More

Tributylborane, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Chemical Identifiers

CAS 122-56-5, 109-99-9
Molecular Formula C12H27B
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009423
InChI Key CMHHITPYCHHOGT-UHFFFAOYSA-N
Synonym tri-n-butylborane, borane, tributyl, tributylborine, tri-n-butyl borane, tributylboron, tri-n-butylboron, ccris 4133, solution, tributyl-borane, tributyl borane
PubChem CID 31216
IUPAC Name tributylborane
SMILES CCCCB(CCCC)CCCC
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

122-56-5, 109-99-9
182.16
CMHHITPYCHHOGT-UHFFFAOYSA-N
31216
CCCCB(CCCC)CCCC
C12H27B
MFCD00009423
tri-n-butylborane, borane, tributyl, tributylborine, tri-n-butyl borane, tributylboron, tri-n-butylboron, ccris 4133, solution, tributyl-borane, tributyl borane
tributylborane

Specifications

Colorless to Yellow
C12H27B
MFCD00009423
10,410
tri-n-butylborane, borane, tributyl, tributylborine, tri-n-butyl borane, tributylboron, tri-n-butylboron, ccris 4133, solution, tributyl-borane, tributyl borane
CCCCB(CCCC)CCCC
182.16
182.16
−20°C
Tributylborane, 1M Solution in Tetrahydrofuran
122-56-5, 109-99-9
[CH3(CH2)3]3B
0.94 to 1.06M
0.8470g/mL
CMHHITPYCHHOGT-UHFFFAOYSA-N
tributylborane
31216
0.847
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Catches fire spontaneously if exposed to air. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. Handle under inert gas. Protect from moisture. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

missing translation for 'einecsNumber' : 204-554-6

missing translation for 'rtecsNumber' : ED1850000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only