missing translation for 'onlineSavingsMsg'
Learn More

Tributylamine, 99%

Product Code. 10133190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
250mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133190 1 L Glass bottle 1 L
10447340 100 mL Glass bottle 100mL
10387053 250 mL Glass bottle 250mL
10377053 2.5 L Glass bottle 2.5 L
11975171 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10133190 Supplier Thermo Scientific Acros Supplier No. 139320010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-82-9 | C12H27N | 185.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-82-9
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.35
MDL Number MFCD00009431
InChI Key IMFACGCPASFAPR-UHFFFAOYSA-N
Synonym tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
PubChem CID 7622
ChEBI CHEBI:38905
IUPAC Name N,N-dibutylbutan-1-amine
SMILES CCCCN(CCCC)CCCC

Specifications

Melting Point -70.0°C
Density 0.7780g/mL
Boiling Point 215.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula [CH3NCH2)3]3N
Refractive Index 1.4273 to 1.4293
Quantity 1 L
Beilstein 04,157
Fieser 01,1189; 06,601
Merck Index 15,978
Specific Gravity 0.778
Solubility Information Solubility in water: 0.08g/L water (20°C). Other solubilities: soluble in most organic solvents,very soluble in alcohol and ether
Viscosity 2.4 mPa.s (20°C)
Formula Weight 185.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tributylamine
Show More Show Less
compliance-icons
Product Identifier
  • Tributylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tributylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.