missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(vinyl)tin, TRC

Product Code. 30457286
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30457286 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30457286

Brand: Toronto Research Chemicals TRCT7738151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tributyl(vinyl)tin
CAS 7486-35-3
Molecular Formula C14 H30 Sn
Quantity 1 g
Synonym Stannane','tributylethenyl- (9CI','ACI), Stannane','tributylvinyl- (8CI), Tin','tributylvinyl- (6CI), Tributylethenylstannane (ACI), (Ethenyl)tributyltin, (Tributylstannyl)ethene, (Tributylstannyl)ethylene, Ethenyltributylstannane, Tributyl(vinyl)stannane, Tributylvinylstannane, Tributylvinyltin, Vinyltributylstannane, Vinyltributyltin
InChI Formula InChI=1S/3C4H9.C2H3.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;1H,2H2;
SMILES CCCC[Sn](CCCC)(CCCC)C=C
IUPAC Name tributyl(ethenyl)stannane
Molecular Weight (g/mol) 317.1
Formula Weight 318.1369
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.