missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Citrate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Tributyl citrate |
| CAS | 77-94-1 |
| Molecular Formula | C18 H32 O7 |
| Quantity | 100 g |
| Synonym | Tributyl 2-Hydroxypropane-1,2,3-tricarboxylate, Tributyl Acetylcitrate Imp. A (EP), Tributyl Citrate, 1,2,3-Propanetricarboxylic acid','2-hydroxy-','tributyl ester (9CI), Citric acid','tributyl ester (6CI,7CI,8CI), 2-Hydroxy-1,2,3-propanetricarboxylic acid tributyl ester, Butyl citrate, Citroflex 4, Citroflex C 4, Citrofol B 1, Citrofol BI, Morflex TBC, NSC 8491, Scandinol SP 21, TBC, Tri-n-butyl citrate, Tributyl citrate, Uniplex 83 |
| InChI Formula | InChI=1S/C18H32O7/c1-4-7-10-23-15(19)13-18(22,17(21)25-12-9-6-3)14-16(20)24-11-8-5-2/h22H,4-14H2,1-3H3 |
| SMILES | CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC |
| IUPAC Name | tributyl 2-hydroxypropane-1,2,3-tricarboxylate |
| Molecular Weight (g/mol) | 360.44 |
| Formula Weight | 360.2148 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction