missing translation for 'onlineSavingsMsg'
Learn More

Tributyl citrate, 99+%

Product Code. 10168230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10168230 1 kg Glass bottle 1kg
10023823 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10168230 Supplier Thermo Scientific Acros Supplier No. 421440010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-94-1 | C18H32O7 | 360.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-94-1
Molecular Formula C18H32O7
Molecular Weight (g/mol) 360.45
MDL Number MFCD00027217
InChI Key ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
Synonym tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
PubChem CID 6507
IUPAC Name tributyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC

Specifications

Acidity 0.02% max. (as citric acid)
Melting Point -20°C
Density 1.0420g/mL
Boiling Point 325°C
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HOCCOO(CH2)3CH3(CH2COO(CH2)3CH3)2
Refractive Index 1.4430 to 1.446
Quantity 1 kg
Merck Index 15, 1565
Specific Gravity 1.042
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic liquids,soluble in ethanol and ether
Viscosity 31.9 mPa.s (25°C)
Formula Weight 360.45
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Tributyl citrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.