missing translation for 'onlineSavingsMsg'
Learn More

Tributyl borate, 98%, AcroSeal™

Product Code. 10566230 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 10566230

Brand: Thermo Scientific Acros 428668000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 688-74-4 | C12H27BO3 | 230.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-74-4
Molecular Formula C12H27BO3
Molecular Weight (g/mol) 230.15
MDL Number MFCD00009434
InChI Key LGQXXHMEBUOXRP-UHFFFAOYSA-N
Synonym n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
PubChem CID 12712
IUPAC Name tributyl borate
SMILES B(OCCCC)(OCCCC)OCCCC

Specifications

Melting Point -70.0°C
Color Colorless
Density 0.8530g/mL
Boiling Point 230.0°C to 235.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3CH2CH2CH2O)3B
Refractive Index 1.4080 to 1.4100
Quantity 800 mL
Specific Gravity 0.853
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with most common organic liquids
Formula Weight 230.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tributyl borate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.