missing translation for 'onlineSavingsMsg'
Learn More

tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane, Thermo Scientific™

Product Code. 10012164
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10012164 1 g Amber Glass Bottle 1g
10205324 250 mg Amber Glass Bottle 250mg
10662865 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10012164 Supplier Thermo Scientific Maybridge Supplier No. CC58241DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1007847-70-2
Molecular Formula C22H39NO2Sn
Molecular Weight (g/mol) 468.269
InChI Key TYKSDTUCXMRYNR-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy-6-tributylstannyl pyridine, tributyl 6-tetrahydropyran-4-yloxy pyrid-2-yl stannane, 2-oxan-4-yl oxy-6-tributylstannyl pyridine
PubChem CID 45594295
IUPAC Name tributyl-[6-(oxan-4-yloxy)pyridin-2-yl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)OC2CCOCC2

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 468.25
Percent Purity 90%
Chemical Name or Material tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.