missing translation for 'onlineSavingsMsg'
Learn More

tributyl(4-methyl-1,3-thiazol-2-yl)stannane, Thermo Scientific™

Product Code. 10537010
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 251635-59-3
Molecular Formula C16H31NSSn
Molecular Weight (g/mol) 388.201
InChI Key PZPGNMUMILSRSX-UHFFFAOYSA-N
Synonym 4-methyl-2-tributylstannyl thiazole, 4-methyl-2-tributylstannyl-1,3-thiazole, 2-tributylstannyl-4-methylthiazole, thiazole, 4-methyl-2-tributylstannyl, 4-methyl-2-tributylstannanyl-thiazole, acmc-209gho, 4-methyl-2-tributylstannylthiazole, 4-methyl-2-tributylstannanylthiazole, 4-methyl-2-tributylstannyl-thiazole, 4-methyl-2-tributylstannanyl-1,3-thiazole
PubChem CID 16427130
IUPAC Name tributyl-(4-methyl-1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC(=CS1)C

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 388.18
Percent Purity 90%
Chemical Name or Material tributyl(4-methyl-1, 3-thiazol-2-yl)stannane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'