missing translation for 'onlineSavingsMsg'
Learn More

tributyl(4-methyl-1,3-thiazol-2-yl)stannane, Thermo Scientific™

Product Code. 10301860
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301860 1 g Amber Glass Bottle 1g
10537010 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301860 Supplier Thermo Scientific Maybridge Supplier No. CC75541DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 251635-59-3
Molecular Formula C16H31NSSn
Molecular Weight (g/mol) 388.201
InChI Key PZPGNMUMILSRSX-UHFFFAOYSA-N
Synonym 4-methyl-2-tributylstannyl thiazole, 4-methyl-2-tributylstannyl-1,3-thiazole, 2-tributylstannyl-4-methylthiazole, thiazole, 4-methyl-2-tributylstannyl, 4-methyl-2-tributylstannanyl-thiazole, acmc-209gho, 4-methyl-2-tributylstannylthiazole, 4-methyl-2-tributylstannanylthiazole, 4-methyl-2-tributylstannyl-thiazole, 4-methyl-2-tributylstannanyl-1,3-thiazole
PubChem CID 16427130
IUPAC Name tributyl-(4-methyl-1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC(=CS1)C

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 388.18
Percent Purity 90%
Chemical Name or Material tributyl(4-methyl-1, 3-thiazol-2-yl)stannane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.