missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl[3-(trifluoromethyl)phenyl]stannane, Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Quantity | 5 g |
Formula Weight | 435.16 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Tributyl[3-(trifluoromethyl)phenyl]stannane |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'