missing translation for 'onlineSavingsMsg'
Learn More

Tribromoethyl alcohol, MedChemExpress

Product Code. 16422339
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422339 Supplier Medchem Express Supplier No. HYB1372
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tribromoethyl alcohol (2,2,2-Tribromoethanol) is used to animals, particularly rodents, before surgery.

Tribromoethyl alcohol (2,2,2-Tribromoethanol) is used to animals, particularly rodents, before surgery.

Specifications

Chemical Name or Material Tribromoethyl alcohol
CAS 75-80-9
Color White
Purity Grade Notes Research
Molecular Formula C2H3Br3O
Quantity 1 g
Synonym 2,2,2-Tribromoethanol Narcolan
Solubility Information Ethanol : 800 mg/mL (2829.25 mM; Need ultrasonic) ∣DMSO : ≥ 2.9 mg/mL (10.26 mM)
SMILES BrC(Br)(Br)CO
Molecular Weight (g/mol) 282.76
Formula Weight 282.76
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.