missing translation for 'onlineSavingsMsg'
Learn More

Tribenzyltin chloride, Thermo Scientific™

Product Code. 11394869
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11394869

Brand: Thermo Scientific Alfa Aesar 071201.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3151-41-5
Molecular Formula C21H21ClSn
Molecular Weight (g/mol) 427.559
MDL Number MFCD00014431
InChI Key PUFMEHZUXSPPAM-UHFFFAOYSA-M
Synonym tribenzyltin chloride, tribenzylchlorotin, chlorotribenzyltin, stannane, chlorotribenzyl, chlorid tribenzylcinicity, stannane, tribenzylchloro, chlorotribenzylstannane, tribenzylstannium chloride, tin, tribenzyl-, chloride, stannane, chlorotris phenylmethyl
PubChem CID 18470
IUPAC Name tribenzyl(chloro)stannane
SMILES C1=CC=C(C=C1)C[Sn](CC2=CC=CC=C2)(CC3=CC=CC=C3)Cl

Specifications

Melting Point 143°C
Quantity 5 g
UN Number UN3146
Sensitivity Moisture sensitive
Formula Weight 427.54
Physical Form Powder
Chemical Name or Material Tribenzyltin chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.