missing translation for 'onlineSavingsMsg'
Learn More

Tribenuron, TRC

Product Code. 30009048
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009048 5 mg 5mg
30006403 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009048 Supplier Toronto Research Chemicals Supplier No. TRCT7675505MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106040-48-6
Molecular Formula C14H15N5O6S
Molecular Weight (g/mol) 381.36
Synonym 2-[[[[(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)methylamino]carbonyl]amino]sulfonyl]-benzoic Acid;
IUPAC Name 2-(N-((4-methoxy-6-methyl-1,3,5-triazin-2-yl)(methyl)carbamoyl)sulfamoyl)benzoic acid
SMILES O=C(O)C1=CC=CC=C1S(=O)(NC(N(C2=NC(OC)=NC(C)=N2)C)=O)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H15N5O6S/c1-8-15-12(17-13(16-8)25-3)19(2)14(22)18-26(23,24)10-7-5-4-6-9(10)11(20)21/h4-7H,1-3H3,(H,18,22)(H,20,21)
Formula Weight 381.07
Chemical Name or Material Tribenuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.