missing translation for 'onlineSavingsMsg'
Learn More

Tribenoside, TRC

Product Code. 30136289
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
100 mg
250 mg
missing translation for 'unitSize':
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30136289 50 mg 50mg
30133952 250 mg 250mg
30006253 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30136289 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT94563050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10310-32-4
Molecular Formula C29H34O6
Molecular Weight (g/mol) 478.58
Synonym Alven, Ba 21401, Ethyl 3,5,6-tris-O-(phenylmethyl)-D-glucofuranoside, Ethyl 3,5,6-tri-O-benzyl-D-glucofuranoside, Flebosan, Glivenol, Glyvenal, Hemocuron, TBGF, Venex
IUPAC Name (3R,4R,5R)-4-(benzyloxy)-5-((R)-1,2-bis(benzyloxy)ethyl)-2-ethoxytetrahydrofuran-3-ol
SMILES CCOC1O[C@@]([C@@H](COCC2=CC=CC=C2)OCC3=CC=CC=C3)([H])[C@H](OCC4=CC=CC=C4)[C@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H34O6/c1-2-32-29-26(30)28(34-20-24-16-10-5-11-17-24)27(35-29)25(33-19-23-14-8-4-9-15-23)21-31-18-22-12-6-3-7-13-22/h3-17,25-30H,2,18-21H2,1H3/t25-,26-,27-,28-,29?/m1/s1
Formula Weight 478.24
Chemical Name or Material Tribenoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.