missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Rabbit anti-Human, Clone: S01-9D1, Novus Biologicals™
Description
TRIB3 Monoclonal antibody specifically detects TRIB3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TRIB3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S01-9D1 |
| Conjugate | Unconjugated |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | C20orf97, dJ1103G7.3, neuronal cell death inducible putative kinase, Neuronal cell death-inducible putative kinase, NIPK, p65-interacting inhibitor of NF-kappaB, p65-interacting inhibitor of NF-kappa-B, SINK, SKIP3, TRB-3, TRB3chromosome 20 open reading frame 97, tribbles homolog 3, tribbles homolog 3 (Drosophila) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human TRIB3 (Uniprot#: Q96RU7) |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?