missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Antibody (1H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057761-M03
This item is not returnable.
View return policy
Description
TRIB3 Monoclonal antibody specifically detects TRIB3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay, Gene Knock-Out, KnockDown
Specifications
| TRIB3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| C20orf97, dJ1103G7.3, neuronal cell death inducible putative kinase, Neuronal cell death-inducible putative kinase, NIPK, p65-interacting inhibitor of NF-kappaB, p65-interacting inhibitor of NF-kappa-B, SINK, SKIP3, TRB-3, TRB3chromosome 20 open reading frame 97, tribbles homolog 3, tribbles homolog 3 (Drosophila) | |
| TRIB3 (AAH27484, 56 a.a. ∽ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PDRATAVATASRLGPYVLLEPEEGGRAYRALHCPTGTEYTCKVYPVQEALAVLEPYARLPPHKHVARPTEVLAGTQLLYAFFTRTHGDMH | |
| 0.1 mg | |
| Apoptosis, Cancer, Hypoxia | |
| 57761 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence, Proximity Ligation Assay, Gene Knock-Out, KnockDown | |
| 1H2 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay, Knockout Validated, Knockdown Validated | |
| AAH27484 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction