missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIB2 Monoclonal Antibody (6A12A12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548735
This item is not returnable.
View return policy
Description
TRIB2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA
This gene encodes one of three members of the Tribbles family. The Tribbles members share a Trb domain, which is homologous to protein serine-threonine kinases, but lacks the active site lysine and probably lacks a catalytic function. The Tribbles proteins interact and modulate the activity of signal transduction pathways in a number of physiological and pathological processes. This Tribbles member induces apoptosis of cells mainly of the hematopoietic origin. It has been identified as a protein up-regulated by inflammatory stimuli in myeloid (THP-1) cells, and also as an oncogene that inactivates the transcription factor C/EBPalpha (CCAAT/enhancer-binding protein alpha) and causes acute myelogenous leukemia. Alternatively spliced transcript variants have been found for this gene.
Specifications
| TRIB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q92519 | |
| Trib2 | |
| Purified recombinant fragment of human TRIB2 (AA: 1-200) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6A12A12 | |
| Unconjugated | |
| Trib2 | |
| AW319517; C5FW; GS3955; RGD1564451; TRB2; TRB-2; trib2; TRIB2 (HGNC:30809); Tribbles homolog 2; tribbles homolog 2 (Drosophila); tribbles pseudokinase 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 28951 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction