missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15694775
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15694775 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694775 Supplier Invitrogen™ Supplier No. PA550851
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total TRIB1 protein.

TRIB1 also called gprotein coupled receptor induced, skip1, gig2, c8fwATP, functions in binding protein binding, protein kinase inhibitor activity, and protein serine/threonine kinase activities. Overexpression of TRIB1 in HeLa cells repressed the basal activity of the IL8 promoter by inhibiting AP1 activity. Overexpression of TRIB1 inhibited oncogenic Ras -driven AP1 activation and MEKK1-mediated AP1 activation. ERK activation was enhanced by TRIB1. Coimmunoprecipitation and yeast 2-hybrid assays showed that MEK1 interacted with both TRIB1 and TRIB3, and MKK4 interacted specifically with TRIB1. Cotransfection of MKK4 enhanced the level of TRIB1, indicating that the TRIB-MAPKK interaction stabilized TRIB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TRIB1
Gene Accession No. Q8K4K4, Q96RU8
Gene Alias A530090O15Rik; C8FW; Gig2; GIG-2; G-protein-coupled receptor induced protein; G-protein-coupled receptor induced protein GIG2; G-protein-coupled receptor-induced gene 2 protein; G-protein-coupled receptor-induced protein 2; phosphoprotein regulated by mitogenic pathways; SKIP1; Trb1; TRB-1; Trib1; tribbles homolog 1; tribbles homolog 1 (Drosophila); tribbles pseudokinase 1; tribbles-like protein 1
Gene Symbols TRIB1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human tribbles pseudokinase 1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10221, 211770
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.