missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triazine & Urea Pesticide Mixture 447 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Atrazine∣Atrazine-desethyl∣Atrazine-desisopropyl∣Bromacil∣Carbetamide∣Chloridazon∣Chlorotoluron∣Chloroxuron∣Chlorpropham∣Cyanazine∣Dimefuron∣Diuron∣Ethidimuron∣Ethofumesate∣Isoproturon∣Linuron∣Metamitron∣Metazachlor∣Methabenzthiazuron∣Metobromuron∣Metolachlor∣Metoxuron∣Monolinuron∣Prometryn∣Propazine∣Simazine∣Terbuthylazine∣Terbuthylazine-desethyl∣Terbutryn |
| CAS | 1912-24-9, 6190-65-4, 1007-28-9, 314-40-9, 16118-49-3, 1698-60-8 |
| Analyte or Component Names | Atrazine∣Atrazine-desethyl∣Atrazine-desisopropyl∣Bromacil∣Carbetamide∣Chloridazon∣Chlorotoluron∣Chloroxuron∣Chlorpropham∣Cyanazine∣Dimefuron∣Diuron∣Ethidimuron∣Ethofumesate∣Isoproturon∣Linuron∣Metamitron∣Metazachlor∣Methabenzthiazuron∣Metobromuron∣Metolachlor∣Metoxuron∣Monolinuron∣Prometryn∣Propazine∣Simazine∣Terbuthylazine∣Terbuthylazine-desethyl∣Terbutryn |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Atrazine 100 μg/mL∣Atrazine-desethyl 100 μg/mL∣Atrazine-desisopropyl 100 μg/mL∣Bromacil 100 μg/mL∣Carbetamide 100 μg/mL∣Chloridazon 100 μg/mL∣Chlorotoluron 100 μg/mL∣Chloroxuron 100 μg/mL∣Chlorpropham 100 μg/mL∣Cyanazine 100 μg/mL∣Dimefuron 100 μg/mL∣Diuron 100 μg/mL∣Ethidimuron 100 μg/mL∣Ethofumesate 100 μg/mL∣Isoproturon 100 μg/mL∣Linuron 100 μg/mL∣Metamitron 100 μg/mL∣Metazachlor 100 μg/mL∣Methabenzthiazuron 100 μg/mL∣Metobromuron 100 μg/mL∣Metolachlor 100 μg/mL∣Metoxuron 100 μg/mL∣Monolinuron 100 μg/mL∣Prometryn 100 μg/mL∣Propazine 100 μg/mL∣Simazine 100 μg/mL∣Terbuthylazine 100 μg/mL∣Terbuthylazine-desethyl 100 μg/mL∣Terbutryn 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction