missing translation for 'onlineSavingsMsg'
Learn More

Triamterene, Mikromol™

Product Code. 17923626
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17923626 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17923626 Supplier Mikromol Supplier No. MM0269.00
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Triamterene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triamterene
CAS 396-01-0
Analyte or Component Names Triamterene
Shipping Condition Room Temperature
Molecular Formula C12 H11 N7
Certifications/Compliance ISO 17025
Synonym 6-Phenyl-2,4,7-pteridinetriamine, 2,4,7-Triamino-6-phenyl-pteridine, 2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-triaminopteridine, Ademin, Ademine, Diren, Ditak, Diurene, Dyren, Dyrenium, Dytac, Jatropur, NSC 77625, Noridil, Pterofen, Pterophene, SKF 8542, Taturil, Teriam, Teridin, Tri-Span, Triampur, Triamteren, Triamterene, Triamteril, Triamteril complex, Triteren, Urocaudal
InChI Formula InChI=1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)
SMILES Nc1nc(N)c2nc(c(N)nc2n1)c3ccccc3
IUPAC Name 6-phenylpteridine-2,4,7-triamine
Molecular Weight (g/mol) 253.26
Quantity 500 mg
Formula Weight 253.1076 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.