missing translation for 'onlineSavingsMsg'
Learn More

Triamterene, MedChemExpress

Product Code. 16452176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452176 500 mg 500mg
16418376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452176 Supplier Medchem Express Supplier No. HYB0575
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Triamterene blocks epithelial Na+ channel (ENaC) in a voltage-dependent manner, which used as a mild diuretic. Triamterene as an inhibitor of the TGR5 receptor.

Triamterene blocks epithelial Na+ channel (ENaC) in a voltage-dependent manner, which used as a mild diuretic. Triamterene as an inhibitor of the TGR5 receptor.

Specifications

Chemical Name or Material Triamterene
Physical Form Solid
CAS 396-01-0
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H11N7
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 4.55 mg/mL (17.97 mM; ultrasonic and adjust pH to 9 with NaOH) ∣H2O : 0.1 mg/mL (0.39 mM; Need ultrasonic)
SMILES NC1=NC(N)=C2N=C(C3=CC=CC=C3)C(N)=NC2=N1
Molecular Weight (g/mol) 253.26
Formula Weight 253.26
Percent Purity 99.48%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.