missing translation for 'onlineSavingsMsg'
Learn More

Triamcinolone Acetonide, Mikromol™

Product Code. 18072090
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 18072090

Brand: Mikromol MM0198.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Triamcinolone Acetonide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triamcinolone acetonide
CAS 76-25-5
Analyte or Component Names Triamcinolone acetonide
Shipping Condition Room Temperature
Molecular Formula C24 H31 F O6
Certifications/Compliance ISO 17034
Synonym Triamcinolone Hexacetonide Imp. A (EP), Triamcinolone Acetonide, Pregna-1,4-diene-3,20-dione, 9-fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11beta,16alpha)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,16alpha,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (6CI,8CI), (11beta,16alpha)-9-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione-16,17-acetonide, 9-Fluoro-11beta,21-dihydroxy-16alpha,17-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,21-dihydroxy-16alpha,17alpha-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 9alpha-Fluoro-11beta,21-dihydroxy-16alpha,17alpha-isopropylidenedioxy-1,4-pregnadiene-3,20-dione, 9alpha-Fluoro-16alpha-hydroxyprednisolone 16alpha,17alpha-acetonide, Adcortyl, Adcortyl A, Aftach, Aristicort, Aristocort A, Aristocort acetonide, Aristoderm, Azmacort, Berlicort, Delphicort, Extracort, Ftorocort, I-Vation, Kenacort 40, Kenacort 80, Kenacort A, Kenacort A 40, Kenalog, Kenalog 10, Kenalog 40, Ledercort Cream, Lichtena, MaQuaid, NSC 21916, Nasacort, Omcilon A, Paralen, Pol'kortolon, Queyanshusong A, Respicort, Rheudenolone, Rineton, Solodelf, TAC 3, TAC 40, Tamceton, Tramacin, Tri-Nasal, Triacet, Triakort, Triam, Triamcinolone 16,17-acetonide, Triamcinolone 16alpha,17alpha-acetonide, Triamcort-Depot, Triamhexal, Triamonide 40, Trianopollon, Tricinolon, Triesence, Trivaris, Trymex, Valon A 10, Vetalog, Vetazine, Volon A, Volonimat, 9-Fluoro-11beta,21-dihydroxy-16alpha,17-(1-methylethylidenedioxy)pregna-1,4-diene-3,20-dione, Fluocinolone Acetonide Impurity H, Triamcinolone Hexacetonide Impurity A
InChI Formula InChI=1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
IUPAC Name 9α-Fluoro-11β,21-dihydroxy-16α,17α-isopropylidenedioxypregna-1,4-diene-3,20-dione
Molecular Weight (g/mol) 434.5
Quantity 250 mg
Formula Weight 434.2105 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.