missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triamcinolone

Product Code. 15791799 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15791799

Brand: Thermo Scientific Chemicals 462240010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-94-7
Molecular Formula C21H27FO6
Molecular Weight (g/mol) 394.43
InChI Key GFNANZIMVAIWHM-OBYCQNJPSA-N
PubChem CID 31307
ChEBI CHEBI:9667

Specifications

Melting Point 262°C to 263°C
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf
Formula Weight 394.43
Chemical Name or Material Triamcinolone
compliance-icons
Product Identifier
  • Triamcinolone
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Triamcinolone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.