missing translation for 'onlineSavingsMsg'
Learn More

Triallyl phosphite, Thermo Scientific Chemicals

Catalog Number 11343357
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
102-84-1
C9H15O3P
202.19
MFCD00014948
KJWHEZXBZQXVSA-UHFFFAOYSA-N
triallyl phosphite, allyl phosphite, triallylphosphite, triallyloxyphosphine, phosphorous acid, tri-2-propenyl ester, allyl phosphite, c3h5o 3p, phosphorous acid, triallyl ester, allyl phosphite 6ci,7ci, phosphorous acid, tri-2-propen-1-yl ester, tris prop-2-en-1-yl phosphite
66893
tris(prop-2-enyl) phosphite
C=CCOP(OCC=C)OCC=C
This item is not returnable. View return policy
102-84-1
C9H15O3P
202.19
MFCD00014948
KJWHEZXBZQXVSA-UHFFFAOYSA-N
triallyl phosphite, allyl phosphite, triallylphosphite, triallyloxyphosphine, phosphorous acid, tri-2-propenyl ester, allyl phosphite, c3h5o 3p, phosphorous acid, triallyl ester, allyl phosphite 6ci,7ci, phosphorous acid, tri-2-propen-1-yl ester, tris prop-2-en-1-yl phosphite
66893
tris(prop-2-enyl) phosphite
C=CCOP(OCC=C)OCC=C

Triallyl phosphite is employed as a radical precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 84°C (12 mmHg)
Flash Point 86°C (187°F)
Odor Strong
Linear Formula (CH2=CHCH2O)3P
Quantity 10 g
UN Number UN3278
Beilstein 1705535
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 202.19
Physical Form Liquid
Chemical Name or Material Triallyl phosphite
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'