missing translation for 'onlineSavingsMsg'
Learn More

Triallyl phosphite

Product Code. 11353357
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353357 50 g 50g
11343357 10 g 10g
11363357 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353357 Supplier Thermo Scientific Alfa Aesar Supplier No. 017484.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-84-1 | C9H15O3P | 202.19 g/mol

Triallyl phosphite is employed as a radical precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-84-1
Molecular Formula C9H15O3P
Molecular Weight (g/mol) 202.19
MDL Number MFCD00014948
InChI Key KJWHEZXBZQXVSA-UHFFFAOYSA-N
Synonym triallyl phosphite, allyl phosphite, triallylphosphite, triallyloxyphosphine, phosphorous acid, tri-2-propenyl ester, allyl phosphite, c3h5o 3p, phosphorous acid, triallyl ester, allyl phosphite 6ci,7ci, phosphorous acid, tri-2-propen-1-yl ester, tris prop-2-en-1-yl phosphite
PubChem CID 66893
IUPAC Name tris(prop-2-enyl) phosphite
SMILES C=CCOP(OCC=C)OCC=C

Specifications

Color Colorless
Boiling Point 84°C (12 mmHg)
Flash Point 86°C (187°F)
Odor Strong
Linear Formula (CH2=CHCH2O)3P
Quantity 50 g
UN Number UN3278
Beilstein 1705535
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 202.19
Physical Form Liquid
Chemical Name or Material Triallyl phosphite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.