missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter), MWCO: 1000 kd

Product Code. p-7184568
missing translation for 'orderingAttributeHoverText'
Diameter (Metric):
10 mm
20 mm
missing translation for 'unitSize'
10m
1m
Each
This item is not returnable. View return policy

Product Code. 11510980

missing translation for 'mfr': Spectrum™ 131486T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • 1m roll of Biotech CE Dialysis Tubing (16mm flatwidth)
  • 2 Universal tubing closures
  • 5 Tubing opening picks

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

Kits are great for comparing membrane MWCO performance to optimize separation or purification. Provides a 1m length of Biotech-Grade CE membrane tubing (16mm flat width), a pair of universal tubing closures and 5 picks for opening membrane tubing. Tubing is wet packaged in 0.05% sodium azide preservative that easily rinses out with water. Nine different MWCOs avilalable: 0.1-0.5 kD, 0.5-1.0 kD, 3.5-5 kD, 8-10 kD, 20 kD, 50 kD, 100 kD, 300 kD and 1000 kD.

  • Rigidly controlled porosity for better selectivity and separations than Standard Grade RC
  • 9 MWCOs available ranging from 100 to 1,000,000 Daltons
  • Ultrapure, no heavy metal or sulfiide impurities
  • Low protein binding
  • No glycerin removal, cleaning or pre-treatment required

Specifications

Product Type Dialysis Trial Kit
MWCO 1000 kDa
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Kit Contents 1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
Quantity Each
For Use With (Application) Ideal for small, critical dialysis applications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.