missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIAD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76592-0.025mg
This item is not returnable.
View return policy
Description
TRIAD3 Polyclonal specifically detects TRIAD3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRIAD3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 6.3.2.-, ring finger protein 216Ubiquitin-conjugating enzyme 7-interacting protein 1, Triad domain-containing protein 3, TRIAD3Zinc finger protein inhibiting NF-kappa-B, U7I1, UBCE7IP1ubiquitin conjugating enzyme 7 interacting protein 1, ZINE3 ubiquitin-protein ligase RNF216 | |
| Rabbit | |
| 95 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| AAP47174 | |
| RNF216 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the amino-terminus of mouse TRIAD3A. The immunogen is located within amino acids 120 - 170 of TRIAD3A. Amino Acid Squence: TESDPLETQNQSSED | |
| Ion exchange chromatography | |
| RUO | |
| 54476 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction