missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacetoxymanganese Dihydrate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Triacetoxymanganese Dihydrate |
| CAS | 19513-05-4 |
| Molecular Formula | C6H9MnO6 . 2(H2O) |
| Quantity | 25 g |
| Synonym | Manganese (III) Acetate Dihydrate, Manganese Triacetate Dihydrate |
| InChI Formula | InChI=1S/3C2H4O2.Mn.2H2O/c3*1-2(3)4;;;/h3*1H3,(H,3,4);;2*1H2/q;;;+3;;/p-3 |
| SMILES | O.O.[Mn+3].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-] |
| IUPAC Name | manganese(3+);triacetate;dihydrate |
| Molecular Weight (g/mol) | 268.1 |
| Formula Weight | 267.9991 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction