missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacetin-d5, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Triacetin-d5 |
| CAS | 159510-46-0 |
| Molecular Formula | C9 D5 H9 O6 |
| Quantity | 250 mg |
| Synonym | 1,2,3-Propane-1,1,2,3,3-d5-triol','triacetate (9CI), 1,2,3-Propane-1,1,2,3,3-d5-triol triacetate, 1,2,3-Propanetriol-1,1,2,3,3-d5 triacetate, Triacetin-d5, Glycerol-d5 triacetate, Triacetylglycerol-d5, Triacetin-D5 (glycerol-D5) |
| InChI Formula | InChI=1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3/i4D2,5D2,9D |
| SMILES | [2H]C([2H])(OC(=O)C)C([2H])(OC(=O)C)C([2H])([2H])OC(=O)C |
| IUPAC Name | (2,3-diacetyloxy-1,1,2,3,3-pentadeuteriopropyl) acetate |
| Molecular Weight (g/mol) | 223.2347 |
| Formula Weight | 223.1104 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction