missing translation for 'onlineSavingsMsg'
Learn More

Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate Hydrobromide, TRC

Product Code. 30450567
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450567 1 g -
30450551 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30450567 Supplier Toronto Research Chemicals Supplier No. TRCT8864601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149353-23-1
Molecular Formula C26H51BrN4O6
Molecular Weight (g/mol) 595.61
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid 1,4,7-Tris(1,1-dimethylethyl) Ester Hydrobromide, 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Tris(1,1-dimethylethyl) Ester Monohydrobromide, Tri-tert-butyl 1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane Hydrobromide
IUPAC Name tert-butyl 2-[4,7-bis[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]-1,4,7,10-tetrazacyclododec-1-yl]acetate;hydrobromide
SMILES Br.CC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCN(CC(=O)OC(C)(C)C)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H50N4O6.BrH/c1-24(2,3)34-21(31)18-28-12-10-27-11-13-29(19-22(32)35-25(4,5)6)15-17-30(16-14-28)20-23(33)36-26(7,8)9;/h27H,10-20H2,1-9H3;1H
Formula Weight 594.2992
Chemical Name or Material Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.