missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-Sodium Orthophosphate Dodecahydrate, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Melting Point | 75°C |
| Color | White |
| pH | 11.8 to 12 |
| Physical Form | Solid |
| Quantity | 250 g |
| Formula Weight | 380.13g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- tri-Sodium orthophosphate dodecahydrate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Tribasic sodium phosphate dodecahydrate,Trisodium phosphate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?