missing translation for 'onlineSavingsMsg'
Learn More

Tri-Salicylic Acid, TRC

Product Code. 30454878
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454878 100 mg 100mg
30452915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454878 Supplier Toronto Research Chemicals Supplier No. TRCT874800100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85531-17-5
Molecular Formula C21 H14 O7
Molecular Weight (g/mol) 378.33
Synonym Benzoic acid, 2-[(2-hydroxybenzoyl)oxy]-, 2-carboxyphenyl ester, Salicylsalicylsalicylic Acid, Trisalicylic Acid
IUPAC Name 2-[2-(2-hydroxybenzoyl)oxybenzoyl]oxybenzoic acid
SMILES OC(=O)c1ccccc1OC(=O)c2ccccc2OC(=O)c3ccccc3O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H14O7/c22-16-10-4-1-7-13(16)20(25)28-18-12-6-3-9-15(18)21(26)27-17-11-5-2-8-14(17)19(23)24/h1-12,22H,(H,23,24)
Formula Weight 378.074
Chemical Name or Material Salicylsalicylsalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.