missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Carbonate, Extra Pure, SLR specified reagent for general laboratory work, Fisher Chemical™

Product Code. 10396380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10396380 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10396380 Supplier Fisher Chemical Supplier No. P/5080/62

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Melting Point 292°C
Color Yellow
pH 8.2
Physical Form Solid
Quantity 2.5 kg
Formula Weight 100.12g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.