missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-Potassium Citrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
£73.70 - £316.00
Chemical Identifiers
CAS | 6100-05-6 |
---|---|
Molecular Formula | C6H7K3O8 |
Molecular Weight (g/mol) | 324.41 |
MDL Number | 150442 |
InChI Key | PJAHUDTUZRZBKM-UHFFFAOYSA-K |
Synonym | potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate |
PubChem CID | 2735208 |
ChEBI | CHEBI:64746 |
IUPAC Name | tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate |
SMILES | C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+] |
Description
Chemical Identifiers
6100-05-6 | |
324.41 | |
PJAHUDTUZRZBKM-UHFFFAOYSA-K | |
2735208 | |
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate |
C6H7K3O8 | |
150442 | |
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate | |
CHEBI:64746 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+] |
Specifications
180°C | |
White | |
Solid | |
150442 | |
PJAHUDTUZRZBKM-UHFFFAOYSA-K | |
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate | |
2735208 | |
324.42g/mol |
6100-05-6 | |
7.5 to 9.0 | |
C6H7K3O8 | |
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+] | |
324.41 | |
CHEBI:64746 | |
Plastic powder jar |