missing translation for 'onlineSavingsMsg'
Learn More

Tri-POC Tenofovir Dimer (>90%), TRC

Product Code. 30417487
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417487 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417487 Supplier Toronto Research Chemicals Supplier No. TRCT0184901MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tri-POC Tenofovir dimer
CAS 1093279-77-6
Molecular Formula C34 H52 N10 O17 P2
Quantity 1 mg
Synonym 2,4,6,8-Tetraoxa-5-phosphanonanedioic acid','5-[[(1R)-2-[6-[[[[9-[(2R)-5-hydroxy-2,11-dimethyl-5-oxido-9-oxo-3,6,8,10-tetraoxa-5-phosphadodec-1-yl]-9H-purin-6-yl]amino]methyl]amino]-9H-purin-9-yl]-1-methylethoxy]methyl]-','1,9-bis(1-methylethyl) ester','5-oxide
InChI Formula InChI=1S/C34H52N10O17P2/c1-21(2)59-32(45)51-16-56-62(48,49)19-54-24(7)9-43-14-41-26-28(37-12-39-30(26)43)35-11-36-29-27-31(40-13-38-29)44(15-42-27)10-25(8)55-20-63(50,57-17-52-33(46)60-22(3)4)58-18-53-34(47)61-23(5)6/h12-15,21-25H,9-11,16-20H2,1-8H3,(H,48,49)(H,35,37,39)(H,36,38,40)/t24-,25-/m1/s1
SMILES CC(C)OC(=O)OCOP(=O)(O)CO[C@H](C)Cn1cnc2c(NCNc3ncnc4c3ncn4C[C@@H](C)OCP(=O)(OCOC(=O)OC(C)C)OCOC(=O)OC(C)C)ncnc12
IUPAC Name [(2R)-1-[6-[[[9-[(2R)-2-[bis(propan-2-yloxycarbonyloxymethoxy)phosphorylmethoxy]propyl]purin-6-yl]amino]methylamino]purin-9-yl]propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphinic acid
Molecular Weight (g/mol) 934.78
Formula Weight 934.2987
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.